Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003406.1 | NP_001003406.1 | 36 | 6640 | ||
NM_021096.3 | NP_066919.2 | 37 | 6745 | |||
XM_017029035.1 | XP_016884524.1 | 27 | 10018 | |||
XM_017029036.1 | XP_016884525.1 | 27 | 10015 | |||
XM_017029037.1 | XP_016884526.1 | 27 | 9980 | |||
GenBank mRNA | AB032946.1 | - | 18 | 3149 | ||
AF142567.1 | - | 37 | 6745 | |||
AF393329.1 | - | 36 | 6640 |