Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005401.4 | NP_005392.2 | 13 | 2104 | ||
XM_017001941.1 | XP_016857430.1 | 14 | 2191 | |||
GenBank mRNA | AK291641.1 | - | 13 | 2036 | ||
BC101754.1 | - | 12 | 1511 | 8069260 | ||
BC104803.1 | - | 12 | 1511 | 8143806 | ||
HQ116786.1 | - | 11 | 1363 | |||
X82676.1 | - | 13 | 1645 |