Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005401.4 | NP_005392.2 | 13 | 2211 | ||
XM_017001941.1 | XP_016857430.1 | 14 | 2298 | |||
GenBank mRNA | AK291641.1 | - | 13 | 2143 | ||
AK298120.1 | - | 12 | 1804 | |||
BC101754.1 | - | 12 | 1618 | 8069260 | ||
BC104803.1 | - | 12 | 1618 | 8143806 | ||
HQ116786.1 | - | 11 | 1470 | |||
X82676.1 | - | 13 | 1752 |