Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243974.1 | NP_001230903.1 | 15 | 1918 | ||
NM_001243975.1 | NP_001230904.1 | 13 | 1861 | |||
NM_005605.4 | NP_005596.2 | 14 | 1891 | |||
XM_017013610.1 | XP_016869099.1 | 14 | 1684 | |||
GenBank mRNA | AK299415.1 | - | 15 | 1752 | ||
AY007249.1 | - | 13 | 1468 | |||
BC004864.2 | - | 14 | 1778 | 3677098 |