Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000068.3 | NP_000059.3 | 3 | 728 | ||
NM_001127221.1 | NP_001120693.1 | 3 | 728 | |||
NM_001127222.1 | NP_001120694.1 | 3 | 728 | |||
NM_001174080.1 | NP_001167551.1 | 3 | 728 | |||
NM_023035.2 | NP_075461.2 | 3 | 728 | |||
GenBank mRNA | AB035726.1 | - | 3 | 527 | ||
AB035727.2 | - | 3 | 492 | |||
AF004883.1 | - | 3 | 728 | |||
AF004884.1 | - | 3 | 728 | |||
FJ040507.1 | - | 3 | 492 | |||
U79666.1 | - | 3 | 709 | |||
X99897.1 | - | 3 | 767 |