Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248011.1 | NP_001234940.1 | 5 | 941 | ||
NM_024047.4 | NP_076952.1 | 6 | 1037 | |||
NM_198038.2 | NP_932155.1 | 6 | 679 | |||
XM_011532032.2 | XP_011530334.1 | 6 | 3216 | |||
GenBank mRNA | AF265341.1 | - | 6 | 678 | ||
AF273028.1 | - | 6 | 1001 | |||
AK027295.1 | - | 6 | 1027 | |||
AK074845.1 | - | 6 | 679 | |||
AK075408.1 | - | 6 | 957 | |||
AK308028.1 | - | 5 | 708 | |||
AY026252.1 | - | 6 | 1032 | |||
AY359123.1 | - | 6 | 731 | |||
BC000542.2 | - | 6 | 797 | 3163944 | ||
BI552154.1 | - | 678 |