Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248011.1 | NP_001234940.1 | 6 | 1040 | ||
NM_024047.4 | NP_076952.1 | 7 | 1136 | |||
NM_198038.2 | NP_932155.1 | 7 | 778 | |||
XM_011532032.2 | XP_011530334.1 | 7 | 3315 | |||
GenBank mRNA | AF265341.1 | - | 7 | 777 | ||
AF273028.1 | - | 7 | 1100 | |||
AK027295.1 | - | 7 | 1126 | |||
AK074845.1 | - | 7 | 778 | |||
AK075408.1 | - | 7 | 1056 | |||
AY026252.1 | - | 7 | 1131 | |||
AY359123.1 | - | 7 | 830 | |||
BC000542.2 | - | 7 | 896 | 3163944 | ||
BI552154.1 | - | 781 |