Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006208.2 | NP_006199.2 | 4 | 486 | ||
GenBank mRNA | AK314870.1 | - | 4 | 313 | ||
BC059375.1 | - | 4 | 486 | 30341060 | ||
D12485.1 | - | 4 | 482 | |||
M57736.1 | - | 4 | 473 |