Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006208.2 | NP_006199.2 | 14 | 1445 | ||
XM_011535896.1 | XP_011534198.1 | 5 | 374 | |||
GenBank mRNA | AK314870.1 | - | 14 | 1272 | ||
BC059375.1 | - | 14 | 1445 | 30341060 | ||
BX537580.1 | - | 6 | 469 | |||
D12485.1 | - | 14 | 1441 | |||
M57736.1 | - | 14 | 1432 |