Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006208.2 | NP_006199.2 | 25 | 2945 | ||
XM_011535896.1 | XP_011534198.1 | 16 | 1874 | |||
GenBank mRNA | BC059375.1 | - | 25 | 2945 | 30341060 | |
D12485.1 | - | 25 | 2941 | |||
M57736.1 | - | 25 | 2932 |