Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317395.1 | NP_001304324.1 | 7 | 2104 | ||
NM_001317399.1 | NP_001304328.1 | 7 | 2032 | |||
NM_031954.4 | NP_114160.1 | 7 | 2101 | |||
NR_133897.1 | 6 | 1931 | ||||
NR_133898.1 | 6 | 1887 | ||||
GenBank mRNA | AB209358.1 | - | 2 | 4646 | ||
AF113208.1 | - | 7 | 1846 | |||
AK027543.1 | - | 7 | 1907 | |||
AK092170.1 | - | 6 | 1720 | |||
AK123733.1 | - | 4 | 2338 | |||
AY205299.1 | - | 7 | 1907 | |||
BC040062.1 | - | 7 | 1854 | 5494016 |