Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297741.1 | NP_001284670.1 | 2 | 240 | ||
NM_001297742.1 | NP_001284671.1 | 2 | 430 | |||
NM_032320.6 | NP_115696.2 | 3 | 398 | |||
XM_017018405.1 | XP_016873894.1 | 2 | 1409 | |||
XM_017018406.1 | XP_016873895.1 | 3 | 388 | |||
XM_017018407.1 | XP_016873896.1 | 3 | 368 | |||
XM_017018408.1 | XP_016873897.1 | 3 | 368 | |||
XM_017018409.1 | XP_016873898.1 | 2 | 235 | |||
XM_017018410.1 | XP_016873899.1 | 2 | 227 | |||
XM_017018411.1 | XP_016873900.1 | 2 | 227 | |||
GenBank mRNA | AF329102.2 | - | 3 | 398 | ||
AK095946.1 | - | 3 | 327 | |||
AK292392.1 | - | 3 | 323 | |||
AK294259.1 | - | 2 | 219 | |||
AK294768.1 | - | 2 | 145 | |||
AK298226.1 | - | 2 | 115 | |||
AK310935.1 | - | 2 | 123 | |||
AK316163.1 | - | 2 | 430 | |||
AL832981.1 | - | 3 | 379 | |||
AY221959.2 | - | 3 | 441 | |||
BC005071.1 | - | 3 | 360 | 3531347 |