Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006240.2 | NP_006231.2 | 10 | 1067 | ||
NM_152224.1 | NP_689410.1 | 10 | 1067 | |||
NM_152226.1 | NP_689412.1 | 10 | 1067 | |||
XM_005274553.2 | XP_005274610.1 | 12 | 1394 | |||
XM_017029612.1 | XP_016885101.1 | 10 | 1205 | |||
GenBank mRNA | AF023455.1 | - | 7 | 668 | ||
AF027977.1 | - | 7 | 820 | |||
AK290463.1 | - | 7 | 722 | |||
AK292235.1 | - | 10 | 1066 | |||
AK295504.1 | - | 6 | 557 | |||
AK300815.1 | - | 6 | 585 | |||
AK315809.1 | - | 7 | 683 | |||
BC036026.1 | - | 10 | 1066 | 5273322 | ||
X97867.1 | - | 10 | 1069 |