Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076780.1 | NP_001070248.1 | 10 | 1689 | ||
NM_052892.3 | NP_443124.3 | 10 | 1689 | |||
NR_126532.1 | 10 | 1689 | ||||
GenBank mRNA | AB067466.1 | - | 2 | 34 | ||
AK098052.1 | - | 4 | 271 | |||
AY164483.1 | - | 10 | 1689 |