Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270476.1 | NP_001257405.1 | 9 | 1200 | ||
NM_006245.3 | NP_006236.1 | 9 | 1183 | |||
NM_180976.2 | NP_851307.1 | 9 | 1087 | |||
NM_180977.2 | NP_851308.1 | 7 | 865 | |||
GenBank mRNA | AB000634.1 | - | 9 | 1018 | ||
AB000635.1 | - | 7 | 712 | |||
AB209861.1 | - | 7 | 762 | |||
AB451342.1 | - | 9 | 1004 | |||
AB451357.1 | - | 9 | 1004 | |||
AK122701.1 | - | 9 | 1077 | |||
AK290604.1 | - | 9 | 996 | |||
AK291797.1 | - | 9 | 1091 | |||
AK299696.1 | - | 9 | 1056 | |||
BC001095.2 | - | 9 | 1067 | 3507280 | ||
BC001175.1 | - | 9 | 994 | 3355395 | ||
BC010692.1 | - | 9 | 1077 | 3853892 | ||
D78360.1 | - | 9 | 985 | |||
L76702.1 | - | 9 | 1191 |