Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316676.1 | NP_001303605.1 | 7 | 824 | ||
NM_001316677.1 | NP_001303606.1 | 5 | 497 | |||
NM_001323354.1 | NP_001310283.1 | 5 | 2217 | |||
NM_001323355.1 | NP_001310284.1 | 5 | 630 | |||
NM_001323356.1 | NP_001310285.1 | 5 | 624 | |||
NM_001323357.1 | NP_001310286.1 | 3 | 300 | |||
NM_006504.5 | NP_006495.1 | 6 | 653 | |||
NM_130435.4 | NP_569119.1 | 3 | 300 | |||
XM_005252691.2 | XP_005252748.1 | 5 | 497 | |||
XM_011539994.2 | XP_011538296.1 | 5 | 615 | |||
XM_011539995.1 | XP_011538297.1 | 5 | 615 | |||
XM_011539996.1 | XP_011538298.1 | 5 | 615 | |||
XM_011539998.1 | XP_011538300.1 | 5 | 615 | |||
XM_017016467.1 | XP_016871956.1 | 5 | 615 | |||
XM_017016468.1 | XP_016871957.1 | 5 | 404 | |||
XM_017016469.1 | XP_016871958.1 | 5 | 615 | |||
GenBank mRNA | AF406557.1 | - | 6 | 588 | ||
AJ315969.1 | - | 3 | 209 | |||
AJ430580.1 | - | 3 | 218 | |||
AK056111.1 | - | 5 | 2217 | |||
AK290414.1 | - | 3 | 236 | |||
AK291828.1 | - | 5 | 497 | |||
AK299498.1 | - | 5 | 466 | |||
BC050062.1 | - | 7 | 732 | 5274802 | ||
BE246451.1 | - | 3 | 281 | |||
BI559814.1 | - | 6 | 660 | |||
BI910569.1 | - | 3 | 263 | |||
BU159075.1 | - | 3 | 262 | |||
BX648180.1 | - | 3 | 306 | |||
DA470230.1 | - | 3 | 300 | |||
X54134.1 | - | 6 | 425 |