Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007026.3 | NP_008957.1 | 3 | 769 | ||
XM_005256977.3 | XP_005257034.1 | 3 | 1863 | |||
XM_011524234.1 | XP_011522536.1 | 3 | 1050 | |||
GenBank mRNA | AF038844.1 | - | 3 | 708 | ||
AF120032.1 | - | 1 | 477 | |||
AK027210.1 | - | 2 | 1458 | |||
AK314894.1 | - | 3 | 718 | |||
BC000370.2 | - | 3 | 680 | 2819474 | ||
BC001894.1 | - | 3 | 710 | 3533971 | ||
BC004448.1 | - | 3 | 680 | 2819474 | ||
CR536500.1 | - | 1 | 475 | |||
CR541800.1 | - | 1 | 475 |