Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144633.2 | NP_653234.2 | 16 | 4633 | ||
GenBank mRNA | AK294417.1 | - | 7 | 3038 | ||
AY053503.1 | - | 16 | 4634 |