Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007207.5 | NP_009138.1 | 4 | 1624 | ||
NR_111939.1 | 3 | 632 | ||||
NR_111940.1 | 3 | 770 | ||||
GenBank mRNA | AB026436.1 | - | 4 | 1669 | ||
AF179212.1 | - | 4 | 1508 | |||
AK022513.1 | - | 3 | 683 | |||
AK293287.1 | - | 5 | 1373 | |||
BC020608.1 | - | 3 | 632 | |||
BC031405.1 | - | 4 | 1523 | 5182027 | ||
BC063826.1 | - | 4 | 1518 | 6178330 | ||
CR457092.1 | - | 3 | 1385 |