Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014214.2 | NP_055029.1 | 8 | 1075 | ||
XM_011525659.2 | XP_011523961.1 | 7 | 857 | |||
XM_011525661.2 | XP_011523963.1 | 8 | 1172 | |||
GenBank mRNA | AF200432.1 | - | 7 | 752 | ||
BC017176.1 | - | 8 | 915 | 3345789 | ||
BT007061.1 | - | 8 | 833 |