Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178527.3 | NP_848622.2 | 3 | 621 | ||
XM_011509426.1 | XP_011507728.1 | 3 | 621 | |||
XM_017001065.1 | XP_016856554.1 | 3 | 316 | |||
XM_017001066.1 | XP_016856555.1 | 3 | 310 | |||
XM_017001067.1 | XP_016856556.1 | 3 | 621 | |||
XM_017001071.1 | XP_016856560.1 | 3 | 228 | |||
GenBank mRNA | AK128104.1 | - | 3 | 621 | ||
BC042592.1 | - | 3 | 568 | 4838901 |