Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020666.2 | NP_065717.1 | 4 | 514 | ||
GenBank mRNA | AF294429.1 | - | 3 | 406 | ||
AK304614.1 | - | 4 | 516 | |||
BC015942.1 | - | 4 | 514 | |||
BC136261.1 | - | 4 | 462 | 9020248 | ||
BC143547.1 | - | 4 | 462 | 9052058 | ||
BC151233.1 | - | 4 | 1728 | |||
CR749504.1 | - | 6 | 1393 |