Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323625.1 | NP_001310554.1 | 6 | 1216 | ||
NM_016361.4 | NP_057445.4 | 6 | 1185 | |||
NR_136633.1 | 6 | 1216 | ||||
NR_136634.1 | 7 | 1591 | ||||
NR_136635.1 | 6 | 1216 | ||||
NR_136636.1 | 6 | 1536 | ||||
XM_011509601.2 | XP_011507903.1 | 6 | 1141 | |||
XM_017001422.1 | XP_016856911.1 | 6 | 1141 | |||
GenBank mRNA | AB030039.2 | - | 6 | 989 | ||
AB031478.1 | - | 6 | 1096 | |||
AB209248.1 | - | 6 | 958 | |||
AK000657.1 | - | 6 | 922 | |||
AK308593.1 | - | 5 | 823 | |||
AK310693.1 | - | 4 | 692 | |||
AY358552.1 | - | 6 | 893 | |||
BC009965.1 | - | 6 | 1165 | 4299943 | ||
BC034686.1 | - | 6 | 1169 | 3463770 | ||
CR457101.1 | - | 6 | 700 | |||
KJ534759.1 | - | 6 | 721 | |||
KJ535036.1 | - | 5 | 593 | |||
KJ535055.1 | - | 6 | 721 |