Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018984.3 | NP_061857.3 | 15 | 3396 | ||
XM_005268984.4 | XP_005269041.1 | 15 | 4107 | |||
XM_005268985.2 | XP_005269042.1 | 14 | 3222 | |||
XM_011538497.1 | XP_011536799.1 | 14 | 3488 | |||
XM_011538499.1 | XP_011536801.1 | 13 | 3287 | |||
XM_011538500.2 | XP_011536802.1 | 14 | 3312 | |||
XM_011538501.2 | XP_011536803.1 | 14 | 3308 | |||
XM_017019491.1 | XP_016874980.1 | 14 | 4334 | |||
XM_017019492.1 | XP_016874981.1 | 15 | 3449 | |||
XM_017019493.1 | XP_016874982.1 | 13 | 3193 | |||
XM_017019494.1 | XP_016874983.1 | 13 | 3288 | |||
GenBank mRNA | AB037719.1 | - | 5 | 2431 | ||
AB072355.1 | - | 15 | 3309 | |||
BC062341.1 | - | 15 | 3388 | 6376684 |