Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286555.1 | NP_001273484.1 | 7 | 3186 | ||
NM_020185.4 | NP_064570.1 | 8 | 1206 | |||
NR_104473.1 | 6 | 3136 | ||||
NR_104474.1 | 6 | 3103 | ||||
NR_104475.1 | 4 | 3019 | ||||
XM_011514757.2 | XP_011513059.1 | 4 | 2648 | |||
XM_017011062.1 | XP_016866551.1 | 5 | 640 | |||
GenBank mRNA | AB208997.1 | - | 4 | 2539 | ||
AF165519.1 | - | 8 | 747 | |||
AF424702.1 | - | 8 | 806 | |||
AK128760.1 | - | 4 | 3507 | |||
AY249859.1 | - | 8 | 1210 | |||
BC016844.1 | - | 3 | 465 | |||
BC022847.2 | - | 8 | 1162 | 3942055 | ||
BX107497.1 | - | 1 | 310 |