Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002848.3 | NP_002839.1 | 16 | 3081 | ||
NM_030667.2 | NP_109592.1 | 16 | 3081 | |||
NM_030668.2 | NP_109593.1 | 4 | 672 | |||
NM_030669.2 | NP_109594.1 | 4 | 672 | |||
NM_030670.2 | NP_109595.1 | 4 | 672 | |||
NM_030671.2 | NP_109596.1 | 4 | 672 | |||
XM_017019725.1 | XP_016875214.1 | 16 | 3053 | |||
GenBank mRNA | AF152378.1 | - | 4 | 664 | ||
AF187042.1 | - | 4 | 664 | |||
AF187043.1 | - | 4 | 664 | |||
AF187044.1 | - | 4 | 664 | |||
AK289784.1 | - | 16 | 2826 | |||
AK290982.1 | - | 4 | 395 | |||
AK299529.1 | - | 1 | 47 | |||
BC126201.1 | - | 16 | 2781 | 8991917 | ||
BC126203.1 | - | 16 | 2781 | 8991919 | ||
U20489.1 | - | 16 | 2783 | |||
Z48541.1 | - | 16 | 2731 |