Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002848.3 | NP_002839.1 | 17, 18 | 3129 | ||
NM_030667.2 | NP_109592.1 | 18, 19 | 3213 | |||
NM_030668.2 | NP_109593.1 | 5, 6 | 720 | |||
NM_030669.2 | NP_109594.1 | 6, 7 | 804 | |||
NM_030670.2 | NP_109595.1 | 5, 6 | 720 | |||
NM_030671.2 | NP_109596.1 | 6, 7 | 804 | |||
XM_017019725.1 | XP_016875214.1 | 17, 18 | 3101 | |||
GenBank mRNA | AF152378.1 | - | 6, 7 | 796 | ||
AF187042.1 | - | 5, 6 | 712 | |||
AF187043.1 | - | 6, 7 | 796 | |||
AF187044.1 | - | 5, 6 | 712 | |||
AK289784.1 | - | 18, 19 | 2958 | |||
AK290982.1 | - | 5, 6 | 443 | |||
AK299529.1 | - | 3, 4 | 179 | |||
BC126201.1 | - | 18, 19 | 2913 | 8991917 | ||
BC126203.1 | - | 18, 19 | 2913 | 8991919 | ||
U20489.1 | - | 17, 18 | 2831 | |||
Z48541.1 | - | 18, 19 | 2863 |