Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267623.1 | NP_001254552.1 | 6 | 915 | ||
NM_001324236.1 | NP_001311165.1 | 7 | 1041 | |||
NM_001324237.1 | NP_001311166.1 | 8 | 946 | |||
NM_001324238.1 | NP_001311167.1 | 7 | 949 | |||
NM_033453.3 | NP_258412.1 | 8 | 1038 | |||
NM_181493.3 | NP_852470.1 | 8 | 987 | |||
NR_052000.1 | 7 | 881 | ||||
NR_052001.2 | 7 | 959 | ||||
NR_052002.1 | 7 | 973 | ||||
XM_006723564.3 | XP_006723627.1 | 9 | 1368 | |||
XM_006723565.3 | XP_006723628.1 | 7 | 1245 | |||
GenBank mRNA | AB062127.1 | - | 8 | 907 | ||
AF026816.2 | - | 8 | 897 | |||
AF063607.1 | - | 8 | 922 | |||
AF219116.1 | - | 8 | 950 | |||
AF334680.1 | - | 7 | 830 | |||
BC010138.2 | - | 8 | 882 | 4124624 |