Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000021.3 | NP_000012.1 | 9 | 1212 | ||
NM_007318.2 | NP_015557.2 | 9 | 1200 | |||
XM_005267864.2 | XP_005267921.1 | 9 | 1206 | |||
XM_005267866.1 | XP_005267923.1 | 9 | 1195 | |||
XM_011536971.2 | XP_011535273.1 | 9 | 1104 | |||
XM_011536972.2 | XP_011535274.1 | 9 | 1468 | |||
XM_011536973.1 | XP_011535275.1 | 9 | 1456 | |||
XM_011536974.1 | XP_011535276.1 | 9 | 1097 | |||
GenBank mRNA | AF416717.1 | - | 7 | 928 | ||
AK122722.1 | - | 7 | 2481 | |||
AK223173.1 | - | 7 | 942 | |||
AK303876.1 | - | 5 | 587 | |||
AK312531.1 | - | 9 | 1196 | |||
BC011729.2 | - | 9 | 1181 | 4423098 | ||
KU178285.1 | - | 7 | 916 | |||
KU178286.1 | - | 7 | 928 | |||
KU178287.1 | - | 2 | 147 | |||
L42110.1 | - | 9 | 1176 | |||
L76517.1 | - | 9 | 1469 | |||
U40379.1 | - | 7 | 916 | |||
U40380.1 | - | 7 | 916 |