Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000396.3 | NP_000387.1 | 3 | 426 | ||
GenBank mRNA | AK130134.1 | - | 3 | 245 | ||
AK312880.1 | - | 3 | 243 | |||
AU118043.1 | - | 3 | 322 | |||
AU119985.1 | - | 3 | 321 | |||
AY429530.1 | - | 4 | 286 | |||
BC016058.1 | - | 3 | 308 | 4860926 | ||
CR541675.1 | - | 2 | 197 | |||
CR541719.1 | - | 2 | 197 | |||
DA251123.1 | - | 3 | 426 | |||
S79895.1 | - | 3 | 338 | |||
U13665.1 | - | 3 | 302 | |||
U20280.1 | - | 3 | 215 |