Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002792.3 | NP_002783.1 | 6 | 798 | ||
GenBank mRNA | AF022815.1 | - | 6 | 667 | ||
AF054185.1 | - | 6 | 728 | |||
AK127210.1 | - | 2 | 3237 | |||
AK312025.1 | - | 6 | 693 | |||
BC001895.1 | - | 6 | 731 | |||
BC004427.1 | - | 6 | 716 | 2819923 | ||
BC014999.1 | - | 6 | 726 | |||
BG575009.1 | - | 6 | 792 | |||
BT007165.1 | - | 6 | 643 | |||
GQ472224.1 | - | 6 | 716 |