Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130725.1 | NP_001124197.1 | 2 | 309 | ||
NM_001144932.2 | NP_001138404.1 | 2 | 662 | |||
NM_002797.4 | NP_002788.1 | 2 | 662 | |||
XM_005267871.2 | XP_005267928.1 | 2 | 657 | |||
GenBank mRNA | AK225888.1 | - | 2 | 427 | ||
AK300714.1 | - | 2 | 407 | |||
AK311895.1 | - | 2 | 418 | |||
BC057840.1 | - | 2 | 417 | 4795732 | ||
BC107720.1 | - | 2 | 413 | 6738409 | ||
BT006777.1 | - | 2 | 233 | |||
BU539357.1 | - | 2 | 285 | |||
BX248299.1 | - | 2 | 394 | |||
BX538001.1 | - | 2 | 409 | |||
CD048996.1 | - | 2 | 341 | |||
S74378.1 | - | 2 | 254 |