Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002813.6 | NP_002804.2 | 2 | 279 | ||
NR_048555.2 | 2 | 279 | ||||
GenBank mRNA | AK315389.1 | - | 2 | 248 | ||
BC002383.2 | - | 2 | 239 | 2961610 | ||
BC004184.2 | - | 2 | 239 | 2961610 | ||
BC004213.2 | - | 2 | 239 | 2961610 | ||
BI832046.1 | - | 2 | 273 | |||
BM850893.1 | - | 2 | 200 | |||
BU956605.1 | - | 2 | 260 | |||
CN260241.1 | - | 2 | 285 |