Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261400.2 | NP_001248329.1 | 4 | 585 | ||
NM_002813.6 | NP_002804.2 | 6 | 900 | |||
NR_048555.2 | 5 | 688 | ||||
GenBank mRNA | AK055339.1 | - | 2 | 1856 | ||
BC002383.2 | - | 6 | 860 | 2961610 | ||
BC004184.2 | - | 6 | 860 | 2961610 | ||
BC004213.2 | - | 6 | 860 | 2961610 |