Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308176.1 | NP_001295105.1 | 13 | 2214 | ||
NM_001792.4 | NP_001783.2 | 14 | 2694 | |||
XM_011525788.1 | XP_011524090.1 | 14 | 2359 | |||
XM_017025514.1 | XP_016881003.1 | 14 | 2665 | |||
GenBank mRNA | AK302831.1 | - | 13 | 2204 | ||
BC036470.1 | - | 14 | 2386 | 5264944 | ||
M34064.1 | - | 11 | 1793 | |||
S42303.1 | - | 14 | 2728 | |||
X54315.1 | - | 14 | 2371 | |||
X57548.1 | - | 11 | 1796 |