Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286457.1 | NP_001273386.1 | 6 | 733 | ||
NM_001286458.1 | NP_001273387.1 | 7 | 908 | |||
NM_001321858.1 | NP_001308787.1 | 6 | 659 | |||
NM_003470.2 | NP_003461.2 | 6 | 864 | |||
NR_135826.1 | 6 | 687 | ||||
XM_017023652.1 | XP_016879141.1 | 6 | 1292 | |||
XM_017023653.1 | XP_016879142.1 | 6 | 728 | |||
GenBank mRNA | AK302481.1 | - | 7 | 834 | ||
AK302771.1 | - | 6 | 665 | |||
AK302872.1 | - | 6 | 687 | |||
AK302912.1 | - | 7 | 852 | |||
AK316441.1 | - | 6 | 734 | |||
AY376241.1 | - | 6 | 695 | |||
Z72499.1 | - | 6 | 864 |