Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286457.1 | NP_001273386.1 | 8, 9 | 967 | ||
NM_001286458.1 | NP_001273387.1 | 9, 10 | 1142 | |||
NM_001321858.1 | NP_001308787.1 | 8, 9 | 893 | |||
NM_003470.2 | NP_003461.2 | 8, 9 | 1098 | |||
NR_135826.1 | 8, 9 | 921 | ||||
XM_017023652.1 | XP_016879141.1 | 8, 9 | 1526 | |||
XM_017023653.1 | XP_016879142.1 | 8, 9 | 962 | |||
GenBank mRNA | AK302481.1 | - | 9, 10 | 1068 | ||
AK302771.1 | - | 8, 9 | 899 | |||
AK302872.1 | - | 8, 9 | 921 | |||
AK302912.1 | - | 9, 10 | 1086 | |||
AK316441.1 | - | 8, 9 | 968 | |||
AY376241.1 | - | 8, 9 | 929 | |||
Z72499.1 | - | 8, 9 | 1098 |