Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003793.3 | NP_003784.2 | 6 | 824 | ||
XM_011545328.2 | XP_011543630.1 | 6 | 574 | |||
GenBank mRNA | AF071748.1 | - | 4 | 388 | ||
AF071749.1 | - | 4 | 290 | |||
AF088886.2 | - | 6 | 824 | |||
AF136279.1 | - | 6 | 823 | |||
AJ007331.1 | - | 6 | 779 | |||
AK313657.1 | - | 6 | 792 | |||
AL137742.1 | - | 4 | 255 | |||
BC011682.2 | - | 6 | 818 | 3535532 | ||
BC036451.1 | - | 6 | 799 | 4830274 | ||
CR541680.1 | - | 4 | 296 |