Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003793.3 | NP_003784.2 | 9 | 1214 | ||
XM_011545328.2 | XP_011543630.1 | 9 | 964 | |||
GenBank mRNA | AF071748.1 | - | 7 | 778 | ||
AF071749.1 | - | 7 | 680 | |||
AF088886.2 | - | 9 | 1214 | |||
AF136279.1 | - | 9 | 1213 | |||
AJ007331.1 | - | 9 | 1169 | |||
AK313657.1 | - | 9 | 1182 | |||
AL137742.1 | - | 7 | 645 | |||
BC011682.2 | - | 9 | 1208 | 3535532 | ||
BC036451.1 | - | 9 | 1189 | 4830274 | ||
CR541680.1 | - | 8 | 686 |