Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003940.2 | NP_003931.2 | 18 | 2324 | ||
XM_011513269.1 | XP_011511571.1 | 17 | 2244 | |||
XM_017007425.1 | XP_016862914.1 | 18 | 2168 | |||
XM_017007426.1 | XP_016862915.1 | 17 | 2135 | |||
XM_017007427.1 | XP_016862916.1 | 15 | 1855 | |||
GenBank mRNA | AK290338.1 | - | 18 | 2271 | ||
AK302404.1 | - | 18 | 2133 | |||
BC016146.1 | - | 18 | 2277 | 3910925 | ||
HM138083.1 | - | 18 | 2243 | |||
U75362.1 | - | 18 | 2337 |