Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243759.1 | NP_001230688.1 | 9 | 966 | ||
NM_004205.4 | NP_004196.4 | 10 | 1781 | |||
NM_171997.2 | NP_741994.1 | 9 | 1046 | |||
XM_005271721.4 | XP_005271778.1 | 10 | 1654 | |||
XM_005271722.2 | XP_005271779.1 | 10 | 1660 | |||
XM_017018539.1 | XP_016874028.1 | 9 | 742 | |||
GenBank mRNA | AF079564.1 | - | 9 | 784 | ||
AF440755.1 | - | 9 | 1046 | |||
AK057225.1 | - | 10 | 1710 | |||
AK292255.1 | - | 9 | 969 | |||
AK301805.1 | - | 9 | 851 | |||
BC002854.1 | - | 10 | 1662 | 3635143 | ||
BC002955.1 | - | 10 | 1662 | 3543435 | ||
BC041366.2 | - | 9 | 951 | 5273400 |