Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300930.1 | NP_001287859.1 | 17, 18 | 2035 | ||
NM_005467.3 | NP_005458.1 | 18, 19 | 2134 | |||
XM_017017043.1 | XP_016872532.1 | 20, 21 | 2300 | |||
XM_017017044.1 | XP_016872533.1 | 19, 20 | 2202 | |||
XM_017017045.1 | XP_016872534.1 | 18, 19 | 2137 | |||
GenBank mRNA | AJ012370.1 | - | 18, 19 | 2070 | ||
AK075390.1 | - | 18, 19 | 2109 | |||
BC096316.3 | - | 18, 19 | 2046 | 40007638 | ||
BC096317.3 | - | 18, 19 | 2045 | 40007639 | ||
BC096318.3 | - | 18, 19 | 2045 | 40007641 |