Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006092.2 | NP_006083.1 | 6 | 2277 | ||
XM_005249568.1 | XP_005249625.1 | 7 | 2332 | |||
XM_005249572.1 | XP_005249629.1 | 5 | 2195 | |||
XM_005249576.1 | XP_005249633.1 | 2 | 1094 | |||
XM_006715633.2 | XP_006715696.1 | 7 | 2369 | |||
XM_011515079.1 | XP_011513381.1 | 8 | 2515 | |||
XM_011515080.2 | XP_011513382.1 | 6 | 2191 | |||
XM_011515081.2 | XP_011513383.1 | 6 | 2341 | |||
XM_011515083.1 | XP_011513385.1 | 7 | 2332 | |||
XM_011515084.1 | XP_011513386.1 | 7 | 2332 | |||
XM_011515085.1 | XP_011513387.1 | 7 | 2332 | |||
XM_011515087.1 | XP_011513389.1 | 7 | 2332 | |||
XM_011515088.2 | XP_011513390.1 | 7 | 2332 | |||
XM_017011674.1 | XP_016867163.1 | 7 | 2332 | |||
XM_017011675.1 | XP_016867164.1 | 6 | 2238 | |||
GenBank mRNA | AF113925.1 | - | 6 | 2178 | ||
AF126484.1 | - | 6 | 2205 | |||
AK023969.1 | - | 6 | 2238 | |||
BC040339.1 | - | 6 | 2192 | 4338930 | ||
KU178514.1 | - | 3 | 1754 |