Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252078.1 | NP_001239007.1 | 15 | 1988 | ||
NM_006313.2 | NP_006304.1 | 14 | 1901 | |||
XM_005269261.2 | XP_005269318.1 | 9 | 1192 | |||
XM_006719718.3 | XP_006719781.1 | 15 | 1986 | |||
XM_017020284.1 | XP_016875773.1 | 13 | 2620 | |||
XM_017020285.1 | XP_016875774.1 | 16 | 3325 | |||
GenBank mRNA | AB011101.2 | - | 14 | 1826 | ||
AF013990.1 | - | 14 | 1784 | |||
AF106069.1 | - | 14 | 1835 | |||
AF153604.1 | - | 15 | 1970 | |||
BC125123.1 | - | 14 | 1835 | 40118994 |