Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006660.4 | NP_006651.2 | 14 | 2259 | ||
NR_133680.1 | 14 | 2213 | ||||
XM_011521164.2 | XP_011519466.1 | 13 | 2201 | |||
GenBank mRNA | AJ006267.3 | - | 14 | 2064 | ||
AK292316.1 | - | 14 | 2185 | |||
BC130373.1 | - | 14 | 2055 | 40146404 | ||
BC136487.1 | - | 14 | 2055 | 9020476 |