Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006785.3 | NP_006776.1 | 17 | 2502 | ||
NM_173844.2 | NP_776216.1 | 16 | 2469 | |||
GenBank mRNA | AB026118.1 | - | 16 | 2276 | ||
AF130356.2 | - | 17 | 2408 | |||
AF316597.1 | - | 17 | 2502 | |||
AK024859.1 | - | 5 | 694 | |||
AK092004.1 | - | 6 | 1826 | |||
AK291386.1 | - | 17 | 2468 | |||
AL137399.1 | - | 3 | 454 | |||
BC030143.2 | - | 16 | 2323 | 4765195 |