Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014464.3 | NP_055279.3 | 9 | 1322 | ||
XM_006715062.2 | XP_006715125.1 | 10 | 1305 | |||
XM_011514497.2 | XP_011512799.1 | 7 | 1154 | |||
XM_017010745.1 | XP_016866234.1 | 9 | 1286 | |||
XM_017010746.1 | XP_016866235.1 | 9 | 1080 | |||
XM_017010747.1 | XP_016866236.1 | 7 | 931 | |||
XM_017010748.1 | XP_016866237.1 | 7 | 760 | |||
GenBank mRNA | AB022277.1 | - | 9 | 1177 | ||
AF195116.1 | - | 9 | 1176 | |||
AF195117.1 | - | 5 | 747 | |||
AK312918.1 | - | 9 | 1286 | |||
BC070278.1 | - | 9 | 1322 | 4607306 |