Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267594.1 | NP_001254523.1 | 6 | 860 | ||
NM_001267595.1 | NP_001254524.1 | 6 | 1115 | |||
NR_051991.1 | 7 | 1301 | ||||
NR_051992.1 | 7 | 1314 | ||||
XM_006719361.3 | XP_006719424.1 | 6 | 995 | |||
XM_006719362.2 | XP_006719425.1 | 7 | 767 | |||
XM_011538244.2 | XP_011536546.1 | 6 | 1035 | |||
XM_011538245.2 | XP_011536547.1 | 5 | 947 | |||
XM_017019229.1 | XP_016874718.1 | 7 | 1133 | |||
XM_017019230.1 | XP_016874719.1 | 7 | 1088 | |||
XM_017019231.1 | XP_016874720.1 | 7 | 1033 | |||
XM_017019232.1 | XP_016874721.1 | 7 | 760 | |||
XM_017019233.1 | XP_016874722.1 | 6 | 1043 | |||
XM_017019234.1 | XP_016874723.1 | 6 | 953 | |||
XM_017019235.1 | XP_016874724.1 | 6 | 686 | |||
XM_017019236.1 | XP_016874725.1 | 6 | 960 | |||
XM_017019237.1 | XP_016874726.1 | 6 | 860 | |||
XM_017019238.1 | XP_016874727.1 | 7 | 1071 | |||
XM_017019239.1 | XP_016874728.1 | 6 | 1111 | |||
XM_017019240.1 | XP_016874729.1 | 7 | 1071 | |||
GenBank mRNA | AF149770.1 | - | 4 | 535 | ||
AK292060.1 | - | 6 | 631 | |||
AK312874.1 | - | 6 | 860 | |||
BC045639.1 | - | 6 | 931 | 5298667 | ||
BX537920.1 | - | 7 | 1314 | |||
BX640784.1 | - | 7 | 1145 |