Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127460.2 | NP_001120932.1 | 26 | 4367 | ||
NM_001166279.1 | NP_001159751.1 | 26 | 4293 | |||
NM_014871.4 | NP_055686.3 | 26 | 4284 | |||
GenBank mRNA | AB014610.2 | - | 26 | 4190 | ||
AK001232.1 | - | 10 | 1552 | |||
AK096679.1 | - | 14 | 2296 | |||
BC024043.1 | - | 26 | 4197 | 3357890 | ||
BC094885.1 | - | 26 | 4364 | 4823602 | ||
BX648106.1 | - | 25 | 4395 |