Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300927.1 | NP_001287856.1 | 4 | 1140 | ||
NM_001308366.1 | NP_001295295.1 | 5 | 1333 | |||
NM_017712.3 | NP_060182.1 | 5 | 1373 | |||
XM_006722783.3 | XP_006722846.1 | 5 | 1273 | |||
XM_011528115.1 | XP_011526417.1 | 4 | 1298 | |||
XM_017026913.1 | XP_016882402.1 | 4 | 1223 | |||
GenBank mRNA | AK000215.1 | - | 5 | 1375 | ||
AK289968.1 | - | 5 | 1399 | |||
AK294497.1 | - | 4 | 1321 |